Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ GC Agar Base (Dehydrated)

Catalog No. OXCM0367B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0367B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0367B Supplier Thermo Scientific™ Supplier No. CM0367B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively isolate Neisseria species with this medium containing meat and plant enzymatic digests.

Selectively isolate Neisseria species with Thermo Scientific™ Oxoid™ GC Agar Base (Dehydrated). GC Agar Base has been formulated to include Special Peptone LP0072 which is a mixture of meat and plant enzymatic digests. The presence of starch in the medium ensures that toxic metabolites produced by Neisseria are absorbed. Phosphate buffers are included to prevent changes in pH due to amine production that would affect the survival of the organism.

GC Agar Base is employed as a basal medium in the preparation of Chocolate Agar, Thayer-Martin Medium, Modified Thayer-Martin Medium, Martin-Lewis Agar, and Transgrow Agar.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description GC Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 13.9L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.