Learn More
Description
Cultivate streptococci, pneumococci, meningococci and other fastidious organisms with this nutritious medium.
Cultivate streptococci, pneumococci, meningococci and other fastidious organisms with Thermo Scientific™ Oxoid™ Heart Infusion Broth (Dehydrated). The medium is highly nutritious and well buffered to support the growth of wide variety of organisms.
Heart Infusion Broth is developed to give the same performance characteristics as Brain Heart Infusion (BHI) Broth. However, as bovine brain is a specified risk material, the exclusion of it from the Heart Infusion Broth means that it carries a lower regulatory burden.
Cultivate a wide variety of organisms with Heart Infusion Broth.
- Developed to give equivalent performance to Brain Heart Infusion Broth while significantly reducing the regulatory burden due to the exclusion of calf brain infusion.
- Addition of anti-microbial agents such as cycloheximide and chloramphenicol make this medium suitable for the cultivation of yeasts and molds.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | Thermo Scientific Oxoid Heart Infusion Broth |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 13.5L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.