Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Iso-Sensitest Broth (Dehydrated)

Catalog No. OXCM0473B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0473B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0473B Supplier Thermo Scientific™ Supplier No. CM0473B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform effortless antimicrobial susceptibility testing using this semi-defined medium specifically designed to give reproducible results.

Perform effortless testing using Thermo Scientific™ Oxoid™ Iso-Sensitest Broth (Dehydrated), a semi-defined medium specifically designed for antimicrobial susceptibility testing (AST). Iso-Sensitest Broth is carefully constructed to give reproducible results. It allows the growth of the majority of microorganisms without the need of further supplementation.

Iso-Sensitest Broth has stability in mineral content and the ability to produce optimum antimicrobial zones of inhibition. The amino-nitrogen base of acid-hydrolysed casein and special peptones has been supplemented with defined growth factors. Careful preparation of the nutrients ensures that trimethoprim and sulphonamide antagonists are at very low levels.

Iso-Sensitest Broth is produced in parallel with Thermo Scientific™ Oxoid™ Iso-Sensitest Agar CM0471. The broth has an identical nutrient formulation without the specifically purified agar. Where studies on antimicrobial susceptibilities are to be made in both broth and agar, this medium is found to be of particular value.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Iso-Sensitest Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 21.4L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.