Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab-Lemco Agar (Dehydrated)

Catalog No. OXCM0017B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0017B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0017B Supplier Thermo Scientific™ Supplier No. CM0017B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Examine water and dairy products with this nutritious, general purpose medium.

Examine water and dairy products with nutritious Thermo Scientific™ Oxoid™ Lab-Lemco Agar (Dehydrated)1,2. It is a general-purpose medium for general bacteriological use, and may also be used as the basis for selective, differential, or enriched media. The absence of sodium chloride in the formulation prevents Proteus mirabilis from swarming colonies.

Use Lab-Lamco Agar as a general purpose medium for bacteriological purposes.

It can also be used as nutrient meat extract agar for general bacteriology and for the preservation of stock cultures.

Not all products are available for sale in all territories. Please inquire. 
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Lab-Lemco Agar (Nutrient Agar, U.S. Formulation)
Form Powder
Product Type Agar
Quantity 500 g
Yield For 21.7L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.