Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab-Lemco Nutrient Broth (Dehydrated)

Catalog No. oxcm0015b
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0015B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0015B Supplier Thermo Scientific™ Supplier No. CM0015B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Examine water, sewage, and dairy products by American standard methods using this nutritious medium.

Examine water, sewage, and dairy products by American standard methods using Thermo Scientific™ Oxoid™ Lab-Lemaco Nutrient Broth (Dehydrated). It is used as a nutrient meat extract broth for general bacteriology.

Lab-Lemco Broth is a general purpose liquid medium used for the examination of water, dairy and food products1,2,3

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Lab-Lemco Broth (Nutrient Agar) (US formulation)
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 62.5L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.