Learn More
Description
Presumptively identify Escherichia coli from food, water and environmental samples with this selective medium.
Presumptively identify Escherichia coli from food, water and environmental samples with Thermo Scientific™ Oxoid™ Lauryl Tryptose Broth w/MUG (Dehydrated). The MUG (4-methylumbelliferyl-ß-D-glucuronide) is cleaved by the enzyme glucuronidase (GUD) produced by Escherichia coli resulting in a fluorescent end-product.
Escherichia coli is a common Gram-negative microorganism which may be present in food and water samples. The presence of E.coli cells may be indicative of fecal contamination, although their ingestion does not necessarily have adverse effects on health. Nevertheless, certain strains are responsible for diarrhoeal disease and can also lead to more serious forms of illness.
Lauryl Tryptose Broth with MUG provides a selective medium which is used for the detection of coliform organisms in water, dairy products and other foods and with the addition of MUG is a Standard Methods medium recommended by FDA/BAM1.
- Selective: the medium is inhibitory to gram-positive bacteria, especially bacilli and fecal Streptococci making the medium selective for Escherichia coli
- Easy-to-read: gas production as a result of lactose fermentation is a presumptive evidence of the presence of Escherichia coli.
- Recommended by FDA/BAM: for the confirmation of Escherichia coli from water, dairy and other food products.
- Recommended by FDA/BAM: for the confirmation of Escherichia coli from shellfish and meat4
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: AFNOR, BSI, ISO. |
| Description | Lauryl Tryptose Broth w/MUG |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 14.0L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.