Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Listeria Selective Supplement

Catalog No. SR0140B
Change view
Click to view available options
Yield:
Each vial for 2.5L medium
Each vial for 500mL medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Yield
SR0140B Each vial for 2.5L medium
OXSR0140E Each vial for 500mL medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SR0140B Supplier Thermo Scientific™ Supplier No. SR0140B
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.

Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.

Add to Listeria Selective Agar Base, Part No. CM0856B
TRUSTED_SUSTAINABILITY

Specifications

Description Listeria Selective Supplement, Oxford
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 2.5L medium
Species Listeria
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.