Learn More
Description
Detect and enumerate coliform organisms in clinical samples with Thermo Scientific™ Oxoid™ MacConkey Agar without Salt (Dehydrated). The differential medium suppresses swarming of Proteus species and is, therefore, recommended for urine examination in the investigation of suspected urinary tract infection (UTI).
MacConkey Agar without Salt has the same formulation as MacConkey Agar except that it does not contain added salt and, therefore, provides a ‘low electrolyte medium’ on which most Proteus species do not spread. The medium has found particular favour for use in the examination of urine samples so that overgrowth of other organisms is prevented.
Specifications
Specifications
| Description | MacConkey Agar w/o Salt (Dehydrated) |
| Form | Powder |
| Product Type | MacConkey Agar |
| Quantity | 2.5 kg |
| Yield | for 53.2L medium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.