Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Malt Extract Broth (Dehydrated)

Catalog No. OXCM0057B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0057B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0057B Supplier Thermo Scientific™ Supplier No. CM0057B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivate molds and yeasts with typical morphology and pigmentation with this acidic medium.

Cultivate molds and yeasts with typical morphology and pigmentation with Thermo Scientific™ Oxoid™ Malt Extract Broth (Dehydrated). Mycological peptone in the medium gives a luxuriant growth of yeasts and molds with typical morphology and pigmentation. Due to the acidic nature of the medium, it inhibits the growth of bacteria resulting in improved isolation of yeasts and molds from materials heavily infected with bacteria.

Malt Extract Broth is a liquid medium recommended for the cultivation of molds and yeasts.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Malt Extract Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 25.0L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.