Learn More
Thermo Scientific™ Oxoid™ Membrane FC Agar
Catalog No. OXMM0747B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
OXMM0747B 500 g
1 options
This item is not returnable. View return policy
Catalog No. OXMM0747B Supplier Thermo Scientific™ Supplier No. MM0747B
Add to Cart
Add to Cart
This item is not returnable. View return policy
Oxoid Membrane FC Agar is used for the enumeration of fecal coliform microorganisms in water.
Oxoid Membrane FC Agar is used for the enumeration of fecal coliform microorganisms in water.
TRUSTED_SUSTAINABILITY
Specifications
Description Membrane Faecal Coliform (FC) Agar
Form Powder
Product Type Agar
Quantity 500 g
For Laboratory Use Only