Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories

Catalog No. OXMB0181A
Change view
Click to view available options
Description:
Microbact Holding Tray 12A/12B
Microbact Holding Tray 24E/12E
Microbact Mineral Oil
Microbact Reagent Indole-Kovacs
Microbact Reagent Nitrate A
Microbact Reagent Nitrate B
Microbact Reagent Set D
Microbact Reagent Spot Indole DMACA
Microbact Reagent TDA
Microbact Reagent VP I
Microbact Reagent VP II
Quantity:
1 Bottle
1 Tray
10 mL/Bottle
6 x 10 mL Bottles
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
OXMB0181A Microbact Reagent VP I 10 mL/Bottle
OXMB0184A Microbact Reagent VP II 10 mL/Bottle
MB5370A Microbact Holding Tray 12A/12B 1 Tray
OXMB0180A Microbact Reagent TDA 10 mL/Bottle
MB1082A Microbact Reagent Set D 6 x 10 mL Bottles
MB0187A Microbact Reagent Nitrate B 10 mL/Bottle
MB5128A Microbact Holding Tray 24E/12E 1 Tray
MB1448A Microbact Reagent Spot Indole DMACA 10 mL/Bottle
MB1093A Microbact Mineral Oil 1 Bottle
MB0209A Microbact Reagent Indole-Kovacs 10 mL/Bottle
MB0186A Microbact Reagent Nitrate A 10 mL/Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. OXMB0181A Supplier Thermo Scientific™ Supplier No. MB0181A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Simplify the detection of Gram-negative bacteria with convenient Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories.

Gram-negative bacteria are of significant concern in both clinical infections and food contamination. Identification of the causative agent is crucial in the clinical setting to identify the cause of infection and to establish an appropriate treatment regime. In the food industry, biochemical identification can help to trace the source of contamination. With these GNB reagents and accessories, Enterobacteriaceae and common miscellaneous Gram-negative bacilli are quickly and accurately identified so that proper, fast treatment for the patient may follow.

  • Fast: More results available overnight
  • Simple: Simple test strip or microplate format
  • Easy to Read and Interpret: Results are clearly visible as distinct color reactions that can be interpreted using the Microbact Identification Package (MB1244A)
  • Comprehensive: Identifies >100 Gram-negative bacteria, more unusual Enterobacteriaceae and oxidase-positive, Gram-negative bacteria
  • Flexible: Use single strips or combine systems for more complex identifications. Ideal for use in clinical or food microbiology laboratories
  • Reliable: A comprehensive range of biochemical tests based on published reference methodologies1-4

Additional items required: Mineral Oil (MB1093A), Oxidase Strips (MB0266A), Microbact Identification Package (Windows™) (MB1244A), Indole Reagent (MB0209A), VPI Reagent (MB0181A), VPII Reagent (MB0184A), TDA Reagent (MB0180A), NIT A Reagent (MB0186A), NIT B Reagent (MB0187A). The 6 reagents above are also available as Reagent Set D (MB1082A).

Not all products are available for sale in all territories. Please inquire.

Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Description Microbact Reagent VP I
Format Bottle
Packaging Type 1 Box
Quantity 10 mL/Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.