Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Milk Agar (Dehydrated)

Catalog No. OXCM0021B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0021B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0021B Supplier Thermo Scientific™ Supplier No. CM0021B

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Determine the viable microflora of dairy and water samples with this medium corresponding to the medium described in Dept. Of Health Memo.139/Foods.

Determine the viable microflora of dairy and water samples with Thermo Scientific™ Oxoid™ Milk Agar (Dehydrated) – a nutrient medium enriched with milk solids. Milk Agar is made to a formula corresponding to the official medium described in Dept. of Health Memo. 139/ Foods1. It is recommended for performing the plate count test on milks, rinse waters, milk products and ice cream, etc.

It also complies with the recommendations of EUROGLACE (EEC Ice cream Industries) submitted to the EEC Commission for the examination of ice cream2.

Liquid milk and other dairy products are highly perishable and can spoil in a few days. The bacteria present in dairy products may cause disease or spoilage. Foodborne illness from dairy product consumption of have been predominantly associated with Salmonella enterica, Listeria monocytogenes, Camplylobacter jejuni and Escherichia coli O157: H7.

Milk Agar is a nutrient medium enriched with milk solids for the determination of the viable micro-flora of dairy and water samples.

  • Corresponds to the official medium described in Dept. Of Health Memo.139/Foods1
  • Recommended for performing the plate count test on milks, rinse waters, milk products and ice cream, etc.
  • Complies with the recommendations of EUROGLACE (EEC Ice cream Industries) submitted to the EEC Commission for the examination of ice cream2

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Milk Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 20.8L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.