Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MLCB Agar (Dehydrated)

Catalog No. OXCM0783B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0783B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0783B Supplier Thermo Scientific™ Supplier No. CM0783B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively isolate salmonellae other than Salmonella Typhi and Salmonella Paratyphi A from feces and food samples with this medium.

Selectively isolate salmonellae other than Salmonella Typhi and Salmonella Paratyphi A from and food samples with Thermo Scientific™ Oxoid™ MLCB Agar (Dehydrated). MLCB Agar is based on the formula of Inoue et al.1 for the selective isolation of salmonellae from feces and foods. Visual detection of very small numbers of hydrogen sulphide-producing strains is easy because of the distinctive colonial appearance.

Salmonella infection (salmonellosis) is a common bacterial disease that affects the intestinal tract. Salmonella bacteria typically live in animal and human intestines and are shed through feces. Humans become infected most frequently through contaminated water or food.

Use MLCB Agar for selective isolation of other than Salmonella typhi and Salmonella paratyphi A from feces and food samples. The medium may be inoculated directly with the specimen or from an enrichment culture.

Salmonellae grow as large purple-black colonies due to hydrogen sulphide production. Mannitol is utilized by the organism and the resultant pH fall initiates lysine decarboxylation which controls further downward pH movement and promotes blackening.

MLCB Agar does not depend on lactose fermentation and is therefore recommended when investigating lactose-fermenting salmonellae (Salmonella arizona).

Atypical Salmonella strains that produce little or no hydrogen sulphide grow as mauve-grey colonies and may develop a central black `bull's eye’.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: NMKL standards.
Description MLCB Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 10.2L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.