Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MRS Broth (Dehydrated)

Catalog No. OXCM0359B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0359B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0359B Supplier Thermo Scientific™ Supplier No. CM0359B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and enumerate lactic acid bacteria from food with this non-selective medium.

Isolate and enumerate lactic acid bacteria from food with Thermo Scientific™ Oxoid™ MRS (DeMan, Rogosa and Sharpe) Broth (Dehydrated). This non-selective medium is designed to support profuse growth of lactic acid bacteria.

Use MRS Broth for enumeration and isolation of mesophilic lactic acid bacteria from food samples. MRS Broth may also be used for tests in the identification of lactobacilli, such as temperature dependence, growth in 4% NaCl, growth in 0.4% Teepol, etc. as recommended by Sharpe, Fryer, and Smith1.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AFNOR, IDF, NMKL.
Description MRS Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 9.6L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.