Learn More
Description
Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Oxoid™ Mueller Hinton Broth (Dehydrated), a medium that can be used in internationally recognized standard procedures.
The major use of Mueller Hinton Broth is for Antimicrobial Susceptibility Testing (AST). It has been produced in parallel with Mueller-Hinton Agar CM0337. Where studies on antibiotic susceptibilities are being undertaken both in broth and agar, it will be found to be of particular value to have media of identical nutrient formulation. Mueller-Hinton Broth is recommended for broth dilution Minimum Inhibitory Concentration (MIC) studies1.
Mueller-Hinton Broth will require supplementation with the divalent cations Mg++ and Ca++ after sterilization2. The CLSI recommend the following cation levels Ca++, 20-25mg/L; Mg++, 10-12.5mg/L.
Lysed horse blood or thymidine phosphorylase may be added to the broth to improve the MIC endpoints of sulphonamides and trimethoprim3.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Certifications/Compliance | Conforms to CLSI standards. |
| Description | Mueller Hinton Broth |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 23.8L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.