Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Mycoplasma Agar Base (Dehydrated)

Catalog No. OXCM0401B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0401B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0401B Supplier Thermo Scientific™ Supplier No. CM0401B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivate and isolate Mycoplasma species with this basal medium when used with any satisfactory supplements

Cultivate and isolate Mycoplasma species with Thermo Scientific™ Oxoid™ Mycoplasma Agar Base (Dehydrated) when used with any satisfactory supplementary factors for the growth of Mycoplasma (PPLO).

For the cultivation of Mycoplasma, the medium and all the supplements should be free of any toxic substances even in small amounts1. Mycoplasma Agar Base is free from such inhibitory or toxic substances. It also contains a special mineral supplement which improves the growth and colony characteristics of mycoplasmas without interfering with the clarity of the medium.

Mycoplasma are a mollicute genus of bacteria that lack a cell wall around their cell membranes. This characteristic makes them naturally resistant to many common antibiotics such as penicillin or other beta-lactam antibiotics that target cell wall synthesis. They can be parasitic or saprotrophic. They are mainly parasites on the surface of human and animal epithelium cells.

Mycoplasma Agar Base is formulated as a basal medium to be enriched with any satisfactory supplementary factors used for the growth of mycoplasmas.

The majority of mycoplasma require media enriched with serum for good growth. Also antibacterial agents are necessary to prevent overgrowth of the slow-growing mycoplasmas by contaminating organisms.

Two supplements, Mycoplasma Supplement-G SR0059 and Mycoplasma Supplement-P SR0060 are developed for the improved growth of mycoplasmas. Mycoplasma Supplement-G SR0059 is a general supplement when added to Mycoplasma Agar Base produces a complete selective medium for the propagation of sterol-requiring Mycoplasma species of the classical type.

Mycoplasma Supplement-P SR0060 is a liquid supplement based on the formulation recommended by the Mycoplasma Reference Laboratory, CPHLS, Colindale, which is used in conjunction with Mycoplasma Agar Base to form a bi-phasic medium for the isolation and preliminary identification of Mycoplasma pneumoniae.

Many species of mycoplasmas are aerobes or facultative anaerobes but some prefer micro-aerophilic conditions with the addition of carbon dioxide, or strict anaerobiosis.

Pathogenic strains grow best at 35°C while saprophytic strains often grow between 22°C and 30°C, T-strains have an optimal temperature of 36°C.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Muller-Kauffmann Tetrathionate Broth Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 30.5L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.