Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nutrient Broth No. 2 (Dehydrated)

Catalog No. OXCM0067B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0067B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0067B Supplier Thermo Scientific™ Supplier No. CM0067B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivate fastidious pathogens and other microorganisms with this nutritious medium.

Cultivate fastidious pathogens and other microorganisms with nutritious Thermo Scientific™ Oxoid™ Nutrient Broth No.2 (Dehydrated). This medium is comparable to a meat infusion and is richer in nutrients than Nutrient Broth (CM0001). It gives good growth from small inocula and is recommended for sterility testing for aerobic organisms. It complies with the recommendations in the British Pharmacopoea1 for the composition of a sterility testing medium for aerobes.

This non-selective medium contains beef extracts combined with peptone, Lab Lemco, and sodium chloride to provide the necessary nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.

The medium is ideally suited for sub-culture, particularly as a secondary growth medium for staphylococci which are to be tested for coagulase production. Nutrient Broth No.2 made up at double strength corresponds to the medium recommended by the British Standards Institution2 for use in the determination of the Rideal-Walker Coefficient of Disinfectants.

Nutrient Broth No.2 is the basal medium for Preston Campylobacter Enrichment Broth3,4.

Not all products are available for sale in all territories. Please inquire.
 

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: NMKL.
Description Nutrient Broth No. 2
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 20.0L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.