Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nutrient Gelatin (Dehydrated)

Catalog No. OXCM0635B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0635B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0635B Supplier Thermo Scientific™ Supplier No. CM0635B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiate microorganisms by their proteolytic effects with this this medium.

Differentiate microorganisms by their proteolytic effects with Thermo Scientific™ Oxoid™ Nutrient Gelatin (Dehydrated). Certain Enterobacteriaceae such as Proteus species and Serratia species have the ability to produce proteolytic enzyme that liquifies the gelatin. Therefore, nutrient gelatin is employed for differentiation of micro-organisms by their proteolytic effects.

Differentiate gelatin liquifying micro-organisms by 20°C plate count method with Nutrient Gelatin Agar.
TRUSTED_SUSTAINABILITY

Specifications

Description Nutrient Gelatin (CM135a)
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 3.9L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.