Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orange Serum Agar (Dehydrated)

Catalog No. OXCM0657B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0657B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0657B Supplier Thermo Scientific™ Supplier No. CM0657B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and enumerate microorganisms capable of surviving citrus products with this medium

Isolate and enumerate spoilage organisms of citrus products with Thermo Scientific™ Oxoid™ Orange Serum Agar (Dehydrated). Orange Serum Agar is developed specifically for the isolation and enumeration of microorganisms that are capable of surviving citrus products.1 The low pH of these products limits the growth of microorganisms to those capable of tolerating the acidic environment.

Organisms known to grow in single strength and concentrated citrus juices are lactic acid and acetic acid bacteria, yeasts, and molds2. Of this group of organisms, lactic acid bacteria have been mainly implicated as the cause of spoilage and these have been identified as Lactobacillus plantarum, Lactobacillus brevis, Leuconostoc mesenteroides and Leuconostoc dextranicum3.

Tryptone in the medium provides amino acids and other complex nitrogenous substances while glucose serves as fermentable carbohydrate and energy source. Yeast extract supplies vitamins which stimulate growth. Orange serum provides an optimal environment for the recovery of acid tolerant microorganisms from citrus fruit products.

In comparative studies carried out by Murdock et al.4 Orange Serum Agar was found to be a superior medium when compared to other media used for the total count and detection of spoilage organisms.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Orange Serum Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 13.5L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.