Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Perfringens TSC Agar Base (Dehydrated)

Catalog No. OXCM0587B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0587B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0587B Supplier Thermo Scientific™ Supplier No. CM0587B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively isolate and presumptively identify Clostridium perfringensin food samples with this basal medium.

Selectively isolate and presumptively identify Clostridium perfringens in food samples with Thermo Scientific™ Oxoid™ Perfringens TSC Agar Base (Dehydrated). The medium can either be used on its own or with selective agents to make Tryptose Sulphite (TS) Agar, Tryptose Sulphite Cycloserine (TSC)1 agar or Shahadi Ferguson Perfringens (SPF)2 agar for the presumptive identification and enumeration of Clostridium perfringens.

Meat peptone, soy peptone and yeast extracts provide essential nutrients and vitamins for the development of clostridia. Sodium metabisulphite and ferric ammonium citrate act as an indicator of sulphide reduction, indicated by black coloured colonies.

Some strains of Clostridium perfringens may produce an opaque zone around the colony due to lecithinase activity, but this is not considered to be universal for all Clostridium perfringens strains after overnight incubation3, and both black, lecithinase positive and black, lecithinase negative colonies should be considered as presumptive Clostridium perfringens on TSC Agar and confirmatory tests carried out.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Perfringens Agar Base (TSC/SFP)
Form Powder
Product Type Agar
Quantity 500 g
Yield For 10.9L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.