Description
Oxoid Phosphate Buffered Saline Tablets (Dulbecco A) is used for preparation of balanced salt solution without calcium and magnesium.
Specifications
Specifications
| Description | Phosphate Buffered Saline Tablets |
| Form | Tablet |
| Product Type | Media Supplement |
| Quantity | Each |
| Yield | Each tab for 100mL |
For Laboratory Use Only