Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Plate Count Agar (Dehydrated)

Catalog No. OXCM0325B
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0325B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0325B Supplier Thermo Scientific™ Supplier No. CM0325B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Determine the plate counts of micro-organisms in food, dairy samples, waste water and other material of sanitary importance with this medium.

Determine the plate counts of microorganisms in food, dairy samples, waste water and other material of sanitary importance with Thermo Scientific™ Oxoid™ Plate Count Agar (Dehydrated). Plate Count Agar was developed by Buchbinder et al1 which is recommended by APHA2, AOAC3 and PHLS4. The quality control of Plate Count Agar (APHA) includes testing in accordance with ISO 11133:2014.5

Plate Count Agar is equivalent to the medium recommended by the APHA1, PHLS 2 and ISO3,4 for the plate count of micro-organisms in food.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: NMKL.
Description Plate Count Agar
Form Powder
Media Type Plate Count Agar
Product Type Nutrient Agar
Quantity 500 g
Yield For 28.6L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.