Learn More
Thermo Scientific™ Potato Dextrose Agar (EP/USP/JP/BP) (Dehydrated)
Shop All Thermo Scientific ProductsDescription
Isolate and enumerate yeasts and molds from dairy and food products1 and prepare Aspergillus niger for the Harmonised Microbial Limit Tests from international pharmacopoeia2,3,4 with Thermo Scientific™ Oxoid™ Potato Dextrose Agar (Dehydrated). This medium is suitable for the detection and enumeration of heat resistant molds in thermally processed fruits and fruit products5.
Pigment production is a critical part of the identification of the fungus. The agar used in the medium is carefully screened to ensure correct pigment production by fungi such as Fusaria spp.
Use Potato Dextrose Agar for detection and enumeration of yeasts and molds in foods.
- Suitable for detection of heat resistant molds in thermally processed fruits and fruit products.
- Suitable medium for the preparation of Aspergillus niger
- Conforms to EP/USP/JP Microbial Limit Test standards2,3,4
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: FDA. |
| Description | Potato Dextrose Agar |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
| Yield | For 12.8L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.