Learn More
Thermo Scientific™ Oxoid™ Ringers Solution Tablets
Catalog No. OXBR0052G Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
OXBR0052G Each
1 options
This item is not returnable. View return policy
Catalog No. OXBR0052G Supplier Thermo Scientific™ Supplier No. BR0052G
Add to Cart
Add to Cart
This item is not returnable. View return policy
Oxoid Ringers Solution Tablets is used for the preparation of isotonic diluents for bacterial cells and bacteriological specimens.
Oxoid Ringers Solution Tablets is used for the preparation of isotonic diluents for bacterial cells and bacteriological specimens.
Each tablet prepares 500mL solution.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference; AFNOR, BSI, IDF, ISO, NMKL.
Description Ringers Solution Tablets
Form Tablet
Product Type Media Supplement
Quantity Each
Yield Each tab for 500mL medium
For Laboratory Use Only