Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Rogosa Agar (Dehydrated)

Catalog No. OXCM0627B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0627B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0627B Supplier Thermo Scientific™ Supplier No. CM0627B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively isolate and enumerate lactobacilli from dairy and other pathological samples with this medium.

Selectively isolate and enumerate Lactobacilli with Thermo Scientific™ Oxoid™ Rogosa Agar (Dehydrated). The medium has given excellent results when used in quantitative and qualitative studies of lactobacilli in feces, saliva and mouth rinses, and in dairy products.

Rogosa Agar, a modification of the medium described by Rogosa et al.1, is a selective medium for the isolation and enumeration of lactobacilli. High acetate concentration and low pH of the medium effectively suppress other bacteria, but also many strains of other lactic acid bacteria. The modification of the pH to 6.2 instead of 5.5 alters the selectivity of the medium for the whole group of lactic acid bacteria2,3.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Rogosa Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 6.1L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.