Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sheep Blood Agar Base (Dehydrated)

Catalog No. OXCM0854B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0854B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0854B Supplier Thermo Scientific™ Supplier No. CM0854B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivate and isolate fastidious microorganisms through improved hemolytic reactions with this medium when used with sheep blood.

Cultivate and isolate fastidious microorganisms from clinical samples through improved hemolytic reactions with sheep blood with Thermo Fisher™ Oxoid™ Sheep Blood Agar Base (Dehydrated) when used with sheep blood. Sheep Blood Agar Base was developed to meet the demand for an especially nutritious blood agar base which would permit the maximum recovery of organisms without interfering with their hemolytic reactions when used with sheep blood.

Sheep Blood Agar Base is based on the formulation of Blood Agar Base No.2 CM0271. Blood Agar Base No.2, when supplemented with sheep blood, was occasionally found to result in a mixed hemolytic reaction (alpha and beta hemolysis) for some Group A streptococci (Streptococcus pyogenes). These mixed hemolytic reactions were due to trace amounts of fermentable carbohydrates in yeast extract and physiological differences of sheep blood when compared to horse blood.1

Having identified the causes of the mixed hemolytic reactions, the Sheep Blood Agar Base is formulated to be compatible with sheep blood. Comparisons with other blood agar bases supplemented with sheep blood have shown that with Sheep Blood Agar Base the growth of many bacteria ― especially the fastidious streptococci ― is considerably improved, and the expected beta hemolytic reaction is achieved with Streptococcus pyogenes.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Sheep Blood Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 12.5L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.