Learn More
Description
Differentiate enteric bacteria on the basis of sulphide production, indole production and motility with Thermo Scientific™ Oxoid™ SIM Medium (Dehydrated). The medium contains sodium thiosulphate and ferrous ammonium sulphate, which together act as indicators for the production of hydrogen sulphide. Tryptone incorporated into the medium is a tryptophan-rich peptone and is degraded by specific bacteria to produce indole. The use of 0.35% agar in the medium results in the production of a semi-solid medium, ideal for the examination of motility.
A motility-indole medium has been found to be helpful in the identification of the Enterobacteriaceae; e.g. in the differentiation of Klebsiella from Enterobacter and Serratia species1. For convenience, these two important tests have been combined with sulphide-production in one tube. The production of hydrogen sulphide is a useful diagnostic test in the identification of enteric bacteria and is helpful in the differentiation between Salmonella and Shigella.
Typical reactions of organisms in SIM medium:
| Organism | Motility | H2S | Indole |
| Escherichia coli ATCC® 25922 | V | - | + |
| Proteus hauseri ATCC® 13315 | + | + | + |
| Shigella sonnei ATCC® 25931 | - | - | - |
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: FDA. |
| Description | SIM Medium |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 16.7L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.