Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sorbitol MacConkey Agar (SMAC) with BCIG (Dehydrated)

Catalog No. OXCM0981B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0981B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0981B Supplier Thermo Scientific™ Supplier No. CM0981B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect Escherichia coli O157 with this selective and differential medium.

Detect Escherichia coli O157 with selective and differential Thermo Scientific™ Oxoid™ Sorbitol MacConkey Agar (SMAC) incorporating the chromogen 5-bromo-4-chloro-3-indolyl-b -D-glucuronide (BCIG) (Dehydrated).

SMAC with BCIG combines two different screening mechanisms for the detection of Escherichia coli O157, the failure to ferment sorbitol and the absence of b-glucuronidase activity. In a study with artificially contaminated meat samples, SMAC with BCIG reduced the number of false suspect Escherichia coli O157 colonies by 36%.1

Enterohemorrhagic strains of Escherichia coli and in particular the serotype O157:H7 have presented an increasing concern among food and clinical microbiologists since their role as a causative agent in two major food related outbreaks of hemorrhagic colitis was established in 1982. Although a variety of serotypes are known to produce verotoxins; a three-year study in England and Wales showed that 99% of isolates from patients suffering diarrhoea, bloody diarrhoea or hemolytic uraemic syndrome (HUS) belonged to serogroup O1571.

  • Selective: selective for Escherichia coli O157
  • Differential: Escherichia coli O157:H7 appear as straw coloured colonies while other coli appear as blue/green coloured colonies

The intestinal tract of ruminants is the prime reservoir of Escherichia coli O157 and other enterohemorrhagic Escherichia coli (EHEC) strains, therefore meats derived from cattle, sheep, goat and deer can be expected to be contaminated. Foods implicated in human illness related to Escherichia coli O157 include meats, dairy products, vegetables, salads, apple juice and water2.

The Sorbitol MacConkey Agar (SMAC) with BCIG formulation includes 5-bromo-4-chloro-3-indolyl- b-D-glucuronide (BCIG) as a substrate for b-glucuronidase, an enzyme which is not usually present in Escherichia coli O157 strains. The non-sorbitol-fermenting and b-glucuronidase-negative Escherichia coli O157 will appear as straw-coloured colonies. Organisms with b-glucuronidase activity will cleave the substrate and release indoxyl (or halogenated indoxyl) which is rapidly oxidized to the insoluble indigo or its analogue, leading to a distinct blue-green coloration of the colonies3.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Sorbitol MacConkey Agar w/BCIG
Form Powder
Product Type MacConkey Agar
Quantity 500 g
Yield for 9.7L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.