Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tergitol-7 Agar (Dehydrated)

Catalog No. OXCM0793B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0793B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0793B Supplier Thermo Scientific™ Supplier No. CM0793B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enumerate and differentiate coliforms from water and food samples with this selective, differential medium when used with TTC Solution Supplement.

Enumerate and differentiate coliforms from water and food samples with selective Thermo Scientific™ Oxoid™ Tergitol-7 Agar (Dehydrated) when used with TTC Solution Supplement (SR0148). Tergitol 7 inhibits Gram-positive organisms allowing superior recovery of coliforms. Fermentation of lactose is seen by a change in color of the pH indicator bromothymol blue. Tri-phenyltetrazolium chloride (TTC) is rapidly reduced to red formazan by most coliform organisms except Escherichia coli and Enterobacter aerogenes allowing easy differentiation.

Tergitol-7 Agar is based on the formulation described by Chapman1 and is recommended for the selective isolation and differentiation of the coliform group.

  • Easy-to-read: easily differentiate Escherichia coli (yellow colonies) and Enterobacter species (greenish/yellow colonies) from coliforms (red colonies).
  • Selective: Tergitol-7 inhibits Gram positive organisms and minimizes the swarming of Proteus, allowing superior recovery of coliforms.
  • Recommended: for examination of foodstuffs for fecal contamination2.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: NMKL, ISO.
Description Tergitol-7 Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 9.2L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.