Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tetrathionate Broth Base (Dehydrated)

Catalog No. OXCM0029B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0029B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0029B Supplier Thermo Scientific™ Supplier No. CM0029B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively enrich and isolate Salmonella typhi and other salmonellae from feces, sewage with this selective medium.

Selectively enrich and isolate Salmonella typhi and other salmonellae from feces and sewage with Thermo Scientific™ Oxoid™ Tetrathionate Broth Base (Dehydrated). Organisms which reduce tetrathionate, such a salmonellae, flourish in the medium while many fecal organisms are inhibited1.

Salmonella infection (salmonellosis) is a common bacterial disease that affects the intestinal tract. Salmonella bacteria typically live in animal and human intestines and are shed through feces. Humans become infected most frequently through contaminated water or food.

Tetrathionate Broth is recommended for the selective enrichment method of isolating Salmonella typhi and other salmonellae from feces, sewage, etc.

The combination of thiosulphate and tetrathionate render selectivity to the medium, suppressing commensal coliform organisms. Organisms which possess the enzyme tetrathionate reductase grow in the medium. Salmonella and Proteus species possess the enzyme; Escherichia coli and shigellae do not. Members of Proteus group reduce tetrathionate and may consequently impair the value of this medium for the isolation of salmonellae; this disadvantage of the medium is largely overcome by the addition of 40μg of novobiocin to each milliliter of the incomplete medium before the addition of iodine2,3.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Tetrathionate Broth Base
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 6.5L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.