Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tryptone Glucose Extract Agar (Dehydrated)

Catalog No. OXCM0127B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0127B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0127B Supplier Thermo Scientific™ Supplier No. CM0127B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enumerate bacteria in water and dairy products and detect thermophilic organisms with this medium.

Enumerate bacteria in water and dairy products and detect thermophilic organisms with Thermo Scientific™ Oxoid™ Tryptone Glucose Extract Agar (Dehydrated). The medium is recommended for the plate count of water and dairy products1,2. It can also be used for the detection of thermophilic bacteria in dairy products.

Use Tryptone Glucose Extract Agar for enumeration of water and dairy products and for the detection of thermophilic organisms.

The Standard Methods Committee of the American Public Health Association recommended that sterile milk should be added to this medium only when the dilution of the original specimen was greater than 1 in 10. The addition of 10mL of a sterile 10% solution of Skim Milk Powder LP0031 per liter is suitable for this purpose.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC.
Description Tryptone Glucose Extract Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 20.8L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.