Learn More
Description
Detect indole producing coliforms using Thermo Scientific™ Oxoid™ Tryptone Water (Dehydrated). The ability of coliforms to break down the amino acid tryptophan with the formulation of indole is an important property which is used for the classification and identification of bacteria1,2. Tryptone Water is a good substrate for the production of indole because of its high content of tryptophan and it is more reliable than Peptone Water for this purpose.
Use Trypotone Water for the detection of indole production by coliforms.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: ISO, NMKL. |
| Description | Tryptone Water |
| Form | Powder |
| Product Type | Pre-enrichment Medium |
| Quantity | 500 g |
| Yield | For 33.3L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.