Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Urea Agar Base (Dehydrated)

Catalog No. oxcm0053b
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0053B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0053B Supplier Thermo Scientific™ Supplier No. CM0053B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Detect rapid urease activity of the Proteae and non-rapid urease activity of some Enterobacteriaceae with this medium.

Detect rapid urease activity of the Proteae and non-rapid urease activity of some Enterobacteriaceae with Thermo Scientific™ Oxoid™ Urea Agar Base (Dehydrated).

Urea Agar Base is recommended for the preparation of Christensen medium1 for the detection of rapid urease activity of the urease-positive Proteae. The urea medium may be used for the detection of urea hydrolysis by some other Enterobacteriaceae but the incubation period is much longer 24-48 hours.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, FDA.
Description Urea Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 20.8L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.