Learn More
Description
Isolate Vancomycin Resistant Enterococci (VRE) and High-Level Aminoglycoside Resistant Enterococci (HLARE) from clinical samples with this medium.
Isolate Vancomycin Resistant Enterococci (VRE) and High-Level Aminoglycoside Resistant Enterococci (HLARE) from clinical samples with Thermo Scientific™ Oxoid™ VRE Agar Base (Dehydrated). The VRE Agar Base can be used with any one of the three antibiotic supplements to selectively isolate antibiotic resistant populations amongst pathogenic Enterococci:
Meropenem Supplement (SR0184) for the suppression of contaminating flora.
Gentamicin Supplement (SR0185) for the selective isolation of HLARE.
Vancomycin Supplement (SR0186) for the selective isolation of VRE.
The proliferation of enterococci, resistant to many commonly used antimicrobials is on the increase1. The recent emergence of VRE is of great concern as enterococci can cause bacteraemia, endocarditis and urinary tract infections.
The use of VRE Agar Base complies with recommendations from the Centre for Disease Control and Prevention (CDC) to detect VRE infection in its early stages2.
Vancomycin resistant enterococci can be isolated directly by inoculation of the clinical sample onto supplemented VRE Agar. VRE Agar Base contains an indicator system to detect the growth of aesculin-hydrolysing organisms. Enterococci produce black zones around the colonies from the formation of black iron phenolic compounds derived from aesculin-hydrolyis products and ferrous iron.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Description | VRE Agar Base |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
| Yield | For 11.7L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.