Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Water Plate Count Agar (Dehydrated)

Catalog No. R661122
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R661122 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R661122 Supplier Thermo Scientific™ Supplier No. CM1012B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Enumerate all culturable microorganisms from water using colony count technique this medium while conforming to ISO 6222: 1999

Enumerate all culturable microorganisms from water using colony count technique with Thermo Scientific™ Oxoid™ Water Plate Count Agar (Dehydrated). This nutritious, non-selective medium conforms to ISO 6222: 1999 (E) Water quality — Enumeration of culturable microorganisms — Colony count by inoculation in a nutrient agar culture medium1.

Waters of all kinds invariably contain a variety of microorganisms derived from various sources, such as soil and vegetation. Estimation of the overall numbers provide useful information for the assessment and surveillance of water quality. Separate counts are usually made of the microorganisms which are able to grow and form colonies on nutrient agar media at 36°C and 22°C.

Colony counts are useful for assessing the integrity of ground water sources and the efficiency of water treatment processes such as coagulation, filtration and disinfection and provide an indication of the cleanliness and integrity of the distribution system. They can also be used to assess the suitability of a water supply for the preparation of food and drink to avoid contaminating the product with spoilage organisms.

The main value of colony counts lies in the detection of changes from expected counts, based on frequent, long term monitoring. Any sudden increase in the count can be an early warning of serious pollution and a call for immediate investigation.1

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: ISO.
Description Water Plate Count Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 20.8L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.