Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Wilkins-Chalgren Anaerobe Agar (Dehydrated)

Catalog No. OXCM0619B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0619B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0619B Supplier Thermo Scientific™ Supplier No. CM0619B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate, cultivate and /or carry out susceptibility testing of a wide variety of anaerobic organisms from clinical samples with this medium.

Isolate, cultivate and /or carry out susceptibility testing of a wide variety of anaerobic organisms from samples with Thermo Scientific™ Oxoid™ Wilkins-Chalgren Anaerobe Agar (Dehydrated). Wilkins and Chalgren1, recognizing the need for standard medium for antimicrobial susceptibility testing of anerobic bacteria, developed this medium which does not require the addition of blood.

Wilkins-Chalgren Anaerobe Agar is recommended for the susceptibility testing of anaerobic bacteria using the Receiver Operating Characteristic (ROC) procedure. It is also recommended for the isolation of anaerobic organisms from clinical specimens. It has shown to function well both in Petri dishes and roll tubes2.

The formulation of Wilkins-Chalgren Anaerobe Agar includes yeast extract to supply vitamins and other growth factors such as purines and pyrimidines that are necessary for good growth of Peptostreptococcus anaerobius and Bacteroides melaninogenicus. Arginine is added to ensure sufficient amino acid is available for the growth of Eubacterium lentum. Pyruvate is present as an energy source for asaccharolytic cocci such as Veillonella. It also acts similarly to catalase and degrades traces of hydrogen peroxide, which may be produced by the action of molecular oxygen on medium constituents and interfere with the metabolism of anaerobes. Haemin is found to be essential for the growth of Bacteroides species and menadione for Bacteroides melaninogenicus.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Wilkins-Chalgren Anaerobe Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 11.6L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.