Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ WL Nutrient Agar

Catalog No. OXCM0309B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0309B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0309B Supplier Thermo Scientific™ Supplier No. CM0309B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and enumerate microbial flora in brewing and fermentation processes with this medium.

Isolate and enumerate microbial flora in brewing and fermentation applications with Thermo Scientific™ Oxoid™ WL Nutrient Agar. The medium is based on that of Green and Gray1 and is suitable to be used as an aide for the determination of the microbiological flora in brewing and fermentation processes.

WL Nutrient Agar is recommended for the determination of the microbial flora in brewing and fermentation processes.

Reliable counts of fermentation process are obtained with the medium at 5.5. Adjustment to pH 6.5 facilitates the counting of bakers’ and distillers’ yeasts.

Cycloheximide (SR0222) may be added to the medium at a final concentration of 4 mg/L, to suppress yeast growth, the medium becomes selective for the bacterial contaminants of yeast cultures.

When making microbial counts with this medium the time and temperature of incubation will vary according to the materials tested and the organisms sought. Temperatures of 25°C are used for brewing materials and 30°C for bakers’ yeasts.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description WL Nutrient Agar
Form Powder
Product Type Nutrient Agar
Quantity 500 g
Yield For 6.7L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.