Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Wort Agar (Dehydrated)

Catalog No. OXCM0247B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0247B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0247B Supplier Thermo Scientific™ Supplier No. CM0247B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivate and enumerate yeasts with this general purpose mycological medium.

Cultivate and enumerate yeasts with general purpose mycological Thermo Scientific™ Oxoid™ Wort Agar (Dehydrated). The medium equivalent to the medium described by Parfitt1 duplicates the acidity of natural wort making it optimal for many yeasts but inhibitory to most bacteria.

Wort Agar is a general purpose mycological medium, equivalent to the medium described by Parfitt1 and especially suitable for the cultivation and enumeration of yeasts. The medium, which duplicates the composition of natural wort, is of an acidity which is optimal for many yeasts but inhibitory to most bacteria. Parfitt investigated the relative merits of wort agar and other media for the count of yeasts and molds in butter, and recommended the use of dehydrated whey, malt or wort agar for the purpose. Scarr2 employed a modified wort agar (’osmophilic agar’) for the examination of sugar products for osmophilic yeasts. Scarr’s technique is also used for the determination of osmophilic yeasts occurring in materials used in the manufacture of soft drinks.

TRUSTED_SUSTAINABILITY

Specifications

Description Wort Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 10.0L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.