Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxolinic acid, 98%

Catalog No. AC431510050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431510050 5 g Glass bottle
AC431510250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC431510050 Supplier Thermo Scientific Chemicals Supplier No. 431510050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 14698-29-4 | C13H11NO5 | 261.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14698-29-4
Molecular Formula C13H11NO5
Molecular Weight (g/mol) 261.23
MDL Number MFCD00056775
InChI Key KYGZCKSPAKDVKC-UHFFFAOYSA-N
Synonym oxolinic acid, nidantin, dioxacin, emyrenil, prodoxal, prodoxol, ossian, gramurin, oksaren, oxolinic
PubChem CID 4628
ChEBI CHEBI:138856
IUPAC Name 5-ethyl-8-oxo-[1,3]dioxolo[4,5-g]quinoline-7-carboxylic acid
SMILES CCN1C=C(C(O)=O)C(=O)C2=CC3=C(OCO3)C=C12

Specifications

Melting Point 314.0°C to 316.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,7044
Formula Weight 261.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Oxolinic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 238-750-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.