Learn More
Cayman Chemical RIzatrIptn-d6benzoate salt 5mg
SDP
Supplier:  Cayman Chemical 333695 MG
Encompass_Preferred
An inhibitor of ferroptosis inhibits glutamate-induced cell death in HT22 cells (IC50 276.2 nM) increases GPX1 levels and GPX activity as well as inhibits glutamate-induced mitochondrial superoxide anion production in HT22 cells at 3 M completely inhibits RSL3-induced ferroptosis in HT22 cells at 62.5-1000 nM
Catalog No. 50-294-8619
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.