Learn More
TARGETMOL CHEMICALS INC OXYBERBERINE 20MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5750
Encompass_Preferred
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Oxyberberine (8-Oxoberberine) is isolated from the methanol extract of A. mexicana. It has been used to study the SW480 human colon cancer cell line. purity: 98%
Catalog No. 50-257-9735
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.