Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxymetazoline Hydrochloride, USP, Spectrum™ Chemical
SDP

Catalog No. 18604935 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-604-935 25 g
18-604-934 5 g
18-604-936 100 g
18-604-937 500 g
18-604-938 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-935 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. O135925GM
Add to Cart
Add to Cart

O1359, 2315-02-8, C16H24N2O·HCl

Spectrum™ Chemical Oxymetazoline Hydrochloride, USP is used for temporary relief of congestion in the nose caused by various conditions including the common cold, sinusitis, hay fever, and allergies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Concentration 98.5 to 101.5 %
CAS 2-8-2315
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.