Learn More
Tocris Bioscience™ Oxyntomodulin (porcine, bovine)
Catalog No. 20941
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
20-941 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 20-941 Supplier Tocris Bioscience™ Supplier No. 2094/1
This item is not returnable. View return policy
Endogenous gut peptide; modulates feeding and metabolism
GLP-1 receptor agonist; endogenous preproglucagon derived peptide; modulates feeding and metabolism
Oxyntomodulin (porcine, bovine) is a GLP-1 receptor agonist. Endogenous preproglucagon-derived neuropeptide that modulates feeding and metabolism. Also secreted by intestinal L-cells. Increases cAMP production and inhibits gastric acid secretion in rat stomach. Also weak glucagon receptor agonist.
Specifications
Quantity 1 mg