Learn More
Bioworld Oxyrase for Agar, 100 mL
Supplier: Bioworld 306200582
Oxyrasefor AgarOxyrase for Agar™ creates an anaerobic environment when added to bacteriological agar medium Oxyrase for Agar™ is for media with a pH range of 6.8 to 8.4. When used with OxyDish™, Oxyrase for Agar™ will also remove oxygen from the trapped air space inside of the sealed OxyDish™
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.