Learn More
Bioworld Oxyrase for Broth, 100 mL
Supplier:  Bioworld 306200592
Encompass_Preferred
Oxyrasefor BrothOxyrase For Broth™ makes growing anaerobes easy. All you have to do is add Oxyrase For Broth™ to your broth medium. Oxyrase For Broth™ makes broth medium anaerobic. No need to boil broth tubes again. Oxyrase For Broth™ is a medium supplement that contains the Oxyrase Enzyme System and a blend of substrates to maximize the Oxyrase activity in virtually any broth medium. Instruction to use: Aseptically add 1ml of Oxyrase for Broth to each 50ml of prepared sterile brothe medium (or, add one drop per ml of broth) and incubate at 37°C. The broth medium will become anaerobic with 30 minutes.
Catalog No. 50-254-347
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.