Learn More
Thermo Scientific™ Oxytetracycline Glucose Yeast Extract Agar Base (Dehydrated)
Shop All Thermo Scientific ProductsDescription
Selectively enumerate yeasts and molds from foodstuffs with Thermo Scientific™ Oxoid™ Oxytetracycline Glucose Yeast Extract Agar Base (Dehydrated). 1,2. The medium is based upon the formulation described by Mossel et al.3 which utilizes oxytetracycline selectivity to provide increased counts of yeasts and molds compared to other media that utilize a low pH as a selective agent. Improve the recovery of physically stressed yeast cells on this neutral pH medium4.
Use Oxytetracycline Glucose Yeast Extract Agar for selection and enumeration of yeasts and molds from foodstuffs.
- Increased colony count of yeasts and molds compared to that of low pH media.
- Improved recovery of physically stressed yeast cells.
The choice of suitable medium for enumeration of yeast and moulds is greatly dependent on the nature of the foodstuffs under investigation and the organisms that occur on them5. Oxytetracycline Glucose Yeast Extract Agar Base remains bacteriostatic when inoculated with not greater than 1mL of a 10-1 dilution of foods and subsequently incubated for not greater than 5 days at 25°C as is the customary practice in food mycology2.
For isolation of psychrotrophic yeasts from chilled proteinaceous foods a combination of oxytetracycline and gentamicin is effective6.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: AFNOR, ISO, NMKL. |
| Description | Oxytetracycline Glucose Yeast Extract Agar Base |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
| Yield | For 13.5L medium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.