Learn More
TOKU-E OXYTETRACYCLINE SOLN 10X1ML
Supplier:  TOKU-E O04910X1ML
Encompass_Preferred
Antibiotic, ReadyMade™, sterile-filtered, Oxytetracycline Hydrochloride, 1 mg/ml in water. Useful in microbiology and plant biology.
Catalog No. NC2402972
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.