Learn More
CPC Scientific H-Cys-Tyr-Ile-Ser-Asn-Cys-Pro-Ile-Gly-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific OXYT004B
Encompass
ONE-LETTER SEQUENCE: CYISNCPIG-NH2 (Cys & Cys bridge)MOLECULAR FORMULA: C41H63N11O12S2MOLECULAR WEIGHT:966.15STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [550-21-0], RESEARCH AREA: Hormonal
Catalog No. 50-211-0051
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.