Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ P-3FAX-Neu5Ac
GSA_VAGREENER_CHOICESDP

Catalog No. 576010
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-601-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 57-601-0 Supplier Tocris Bioscience™ Supplier No. 5760/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sialic acid analog; sialyltransferase inhibitor

P-3FAX-Neu5Ac is a cell-permeable sialic acid analog. Deacetylation occurs intracellularly giving rise to a sialyltransferase inhibitor. Abolishes expression of SLex expression on HL-60 cells and reduces E-selectin and P-selectin binding. Prevents metastasis formation in a mouse lung metastasis model.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Chemical Name or Material 5-(acetylamino)-3,5-dideoxy-3-fluoro-D-erythro-α-L-manno-2-nonulopyranosonic acid methyl ester 2,4,7,8,9-pentaacetate
CAS 117405-58-0
Quantity 10 mg
Target Other Transferase Inhibitors
Molecular Formula C22H30FNO14
Formula Weight 551.47
Purity >97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.