Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Anisidine, 99%

Catalog No. p-7081126
Change view
Click to view available options
Quantity:
100 g
500 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1094622 100 g
AAA1094636 500 g
AAA109460B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1094622 Supplier Thermo Scientific Chemicals Supplier No. A1094622
Only null left
Add to Cart

CAS: 104-94-9 | C7H9NO | 123.155 g/mol

p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.

Solubility
Soluble in acetone, benzene, ether and ethanol. Insoluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-94-9
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007864
InChI Key BHAAPTBBJKJZER-UHFFFAOYSA-N
Synonym p-anisidine, 4-anisidine, 4-methoxybenzenamine, 4-aminoanisole, p-aminoanisole, para-anisidine, p-methoxyaniline, benzenamine, 4-methoxy, 4-methoxybenzeneamine, p-anisylamine
PubChem CID 7732
ChEBI CHEBI:82388
IUPAC Name 4-methoxyaniline
SMILES COC1=CC=C(C=C1)N

Specifications

Melting Point 56°C to 60°C
Density 1.089
Boiling Point 240°C to 243°C
Flash Point 122°C (251°F)
Quantity 100 g
UN Number UN2811
Beilstein 471556
Merck Index 14,667
Solubility Information Soluble in acetone,benzene,ether and ethanol. Insoluble in water.
Formula Weight 123.16
Percent Purity 99%
Chemical Name or Material p-Anisidine
Show More Show Less
Hazard Category H300+H310+H330-H350-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P262-P264b-P270-P271-P281-P284-P301+P310-P302+P350-P304+P340-P308+P313-P310-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 203-254-2
RTECSNumber BZ5450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.